missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£361.00
Specifications
| Antigen | GLUT12 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18293072
|
Novus Biologicals
NBP1-60023 |
100 μL | |||||||
Description
GLUT12 Polyclonal specifically detects GLUT12 in Human samples. It is validated for Western Blot.Specifications
| GLUT12 | |
| Polyclonal | |
| Rabbit | |
| Q8TD20 | |
| 154091 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| GLUT-12, GLUT12GLUT8Glucose transporter type 12, solute carrier family 2 (facilitated glucose transporter), member 12, solute carrier family 2, facilitated glucose transporter member 12 | |
| SLC2A12 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title