missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR7/GRIK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £397.00
Specifications
| Antigen | GluR7/GRIK3 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18278064
|
Novus Biologicals
NBP1-46486-0.025ML |
0.025 mL |
£153.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18231351
|
Novus Biologicals
NBP1-46486 |
0.1 mL |
£397.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GluR7/GRIK3 Polyclonal specifically detects GluR7/GRIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| GluR7/GRIK3 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neurotransmission | |
| 2899 | |
| IgG | |
| Unpurified | |
| Specific for GRIK3. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Rat, Mouse, Human | |
| GRIK3 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title