missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR7/GRIK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46486
This item is not returnable.
View return policy
Description
GluR7/GRIK3 Polyclonal specifically detects GluR7/GRIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| GluR7/GRIK3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| GRIK3 | |
| A synthetic peptide from c-terminal region of human GluR7/GRIK3 conjugated to blue carrier protein was used as the antigen. | |
| 0.1 mL | |
| Neuronal Cell Markers, Neurotransmission | |
| 2899 | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000 | |
| EAA5, Excitatory amino acid receptor 5, GLR7, GluR7, GluR-7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for GRIK3. | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction