Learn More
Abcam Glucose-6-Phosphate Assay Kit - High Sensitivity (Fluorometric)
Product Code. 32116718 Shop All Abcam Products
Change view
Click to view available options
100 Tests
Product Code.
32116718 100 Tests
1 options
Product Code. 32116718 Supplier Abcam Supplier No. ab107923100Test
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure 6-O-phosphono-alpha-D-glucopyranose in 20m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with Glucose-6-Phosphate Assay Kit AB107923. Sensitivity>1 μM. Cited in 3 publications. Use with Microplate reader.
Measure 6-O-phosphono-alpha-D-glucopyranose in 20m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with Glucose-6-Phosphate Assay Kit AB107923. Sensitivity>1 μM. Cited in 3 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests