missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLTSCR1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-09493-25UL
This item is not returnable.
View return policy
Description
GLTSCR1 Polyclonal specifically detects GLTSCR1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| GLTSCR1 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| Glioma Tumor Suppressor Candidate Region Gene 1, Glioma Tumor Suppressor Candidate Region Gene 1 Protein | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse Gltscr1 (NP_001074887). Peptide sequence FIQEEKTTLALDKQLAKEKPDEYVSSSRSLGFPVPVSSEGHRLPSHGQSS | |
| 25 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29998 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction