Learn More
STEMCELL Technologies GloCell™ Fixable Viability Dye Violet 540
Product Code. 17119291
Encompass_Preferred
Click to view available options
Quantity:
100 Tests
5 x 100 Tests
Unit Size:
100 tests
Pack of 5
Product Code. 17119291
Brand: STEMCELL Technologies 75011
Amine-labeling fluorescent dye for live/dead staining of mammalian cells
  • GloCell™ Fixable Viability Dye Violet 540 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry
  • The dye irreversibly binds intracellular and cell surface amine groups and can be utilized prior to fixation, permeabilization, or cryopreservation
  • Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay
  • GloCell™ Fixable Viability Dye Violet 540 is excited by the violet laser at 405 nm and it has a fluorescence emission maximum of 540 nm that can be detected using the 525/50 band pass filter used to detect AmCyan
Specifications
For Use With (Application) Flow Cytometry; Immunofluorescence
Quantity 5 x 100 Tests