missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GliS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £423.00
Specifications
| Antigen | GliS2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18675027
|
Novus Biologicals
NBP2-41311-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18115389
|
Novus Biologicals
NBP2-41311 |
0.1 mg |
£423.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GliS2 Polyclonal specifically detects GliS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GliS2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| GLIS family zinc finger 2, NKL, zinc finger protein GLIS2 | |
| GLIS2 | |
| IgG | |
| Affinity Purified | |
| 60 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 84662 | |
| Antibody was raised against an 18 amino acid peptide near the center of human GLIS2. The immunogen is located within amino acids 260 - 310 of GLIS2. Amino Acid Squence: NKRYSNSSDRFKHTRTH | |
| Primary | |
| GLIS2 antibody is human, mouse and rat reactive. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title