missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GliS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41311-0.025mg
This item is not returnable.
View return policy
Description
GliS2 Polyclonal specifically detects GliS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GliS2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| GLIS2 | |
| Antibody was raised against an 18 amino acid peptide near the center of human GLIS2. The immunogen is located within amino acids 260 - 310 of GLIS2. Amino Acid Squence: NKRYSNSSDRFKHTRTH | |
| Affinity Purified | |
| RUO | |
| 84662 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 | |
| GLIS family zinc finger 2, NKL, zinc finger protein GLIS2 | |
| Rabbit | |
| 60 kDa | |
| 0.025 mg | |
| Primary | |
| GLIS2 antibody is human, mouse and rat reactive. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction