missing translation for 'onlineSavingsMsg'
Learn More

PPAR alpha/NR1C1 Antibody - N-terminal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 16 publications

£207.00 - £587.00

Specifications

Antigen PPAR alpha/NR1C1
Concentration 1 mg/ml
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:75000-1:125000, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1-5ug/ml, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18091816
View Documents
Novus Biologicals
NB600-636SS
0.025 mg
£207.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18786083
View Documents
Novus Biologicals
NB600-636
0.1 mg
£587.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PPAR alpha/NR1C1 Polyclonal specifically detects PPAR alpha/NR1C1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PPAR alpha/NR1C1
Western Blot 1:500-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:75000-1:125000, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1-5ug/ml, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Polyclonal
Rabbit
Cancer, mTOR Pathway
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452
PPARA
IgG
Affinity Purified
No reactivity is expected against other subtypes of PPAR.
1 mg/ml
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Unconjugated
RUO
Human, Mouse, Rat, Canine, Hamster, Hamster
P23204
5465
PPAR alpha/NR1C1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a N-Terminal region near amino acids 1-25 of mouse PPAR alpha/NR1C1. (Uniprot: P23204)
Primary
Store at -20°C. Avoid freeze-thaw cycles.
52.35 kDa
Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.