Learn More
PPAR alpha/NR1C1 Antibody - N-terminal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 16 publications
Brand:  Novus Biologicals NB600-636
 View more versions of this product
Product Code. 18786083
  • £587.00 / 0.10mg
Description
PPAR alpha/NR1C1 Polyclonal specifically detects PPAR alpha/NR1C1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PPAR alpha/NR1C1
Polyclonal
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452
Rabbit
52.35 kDa
0.1 mg
Cancer, mTOR Pathway
5465
Human, Mouse, Rat, Canine, Hamster, Hamster
IgG
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
1 mg/ml
Western Blot 1:500-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:75000-1:125000, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1-5ug/ml, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
P23204
PPARA
PPAR alpha/NR1C1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a N-Terminal region near amino acids 1-25 of mouse PPAR alpha/NR1C1. (Uniprot: P23204)
Affinity Purified
RUO
Primary
No reactivity is expected against other subtypes of PPAR.
Store at -20°C. Avoid freeze-thaw cycles.