Learn More
Sperian™ Glasses
Product Code. 11837070
Change view
Click to view available options
:
Product Type: Glasses; Lens Coating: Anti-Fog Coating
Product Code.
11837070 Product Type: Glasses; Lens Coating: Anti-Fog Coating
1 options
Product Code. 11837070 Supplier Sperian™ Supplier No. 1017086
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1775, EC Attestation Number EC1775
Lens Coating Anti-fog, Impact-resistant
Product Type Safety glasses
For Use With (Application) Indoor use. Automotive, Building, Construction, Iron, Steel, Maintenance, Ship Building, Textile, Transportation and Utility industries.
Frame Color Black, Silver
No. per Pack 1 Ea.
Lens Material Polycarbonate
Frame Material Polyamide