Learn More
DWK Life Sciences Glass Stopper, Clear, SBW Glass, Short Standard Taper Ground Joint, Octagonal flat head
Product Code. 15251989 Shop All DWK Life Sciences Products
Click to view available options
Standard Taper Joint:
ST 14/15
ST 24/20
ST 34/24
Unit Size:
Each
Pack of 10
Product Code. 15251989
Brand: DWK Life Sciences 246250802
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Clear SBW glass ground stopper, short-length
Solid or semi-hollow, short-length, clear SBW glass stoppers can be used to close glassware items such as flasks or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head.
  • All glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Octagonal shaped head
TRUSTED_SUSTAINABILITY
Specifications
Description Solid
For Use With (Application) Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders
Quantity 10/Pk.
Standard Taper Joint ST 24/20
Autoclavable Not Autoclavable
Material SBW Glass