Learn More
Duran™ Glass Stoppers
Shop All DWK Life Sciences Products
Click to view available options
Color:
Amber
Unit Size:
Pack of 10
Description
- DURAN™ cap
- Flat head
- Semi-hollow
- Made of Borosilicate glass
Specifications
Specifications
| Quantity | 10/Pk. |
| Standard Taper Joint | NS 14/15 |
| Color | Amber |
| Material | Glass |
| Product Type | Octagonal Head Stopper |
| Shape | Octagonal Grip |