Learn More
DWK Life Sciences™ Safety Stopcock, NS 24/29, for safety outlets type Wertex
Product Code. 10241511 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Borosilicate Glass 3.3, Polypropylene
Unit Size:
Each
Product Code. Material unitSize
10241511 Borosilicate Glass 3.3, Polypropylene Each
1 options
Product Code. 10241511 Supplier DWK Life Sciences™ Supplier No. 247960304
Only null left
Add to basket
Add to basket
The safety stopcock is used for airtight sealing of a desiccator.
The safety stopcock is used for airtight sealing of a desiccator. The stopcock can be firmly connected to the desiccator by means of the matching washer blade.
  • Tight closure due to standard ground joint
  • The safety stopcock can be permanently mounted by means of a sheath
  • For safety outlets of WERTEX type desiccator
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass 3.3, Polypropylene
For Use With (Application) Closure of desiccators
Height (Metric) 76 mm