Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
- With short and wide stem
- Large stem diameter makes it suitable for transferring solids
- Fired-on, highly durable white print
Specifications
Specifications
| Diameter (Metric)Top | 160 mm |
| Material | Borosilicate Glass (3.3) |
| Autoclavable | Autoclavable |
| For Use With (Application) | Transferring of powdered substances and granulated material; Spillage of substances is prevented; Transferring |
| Quantity | 1 Ea. |