Learn More
DWK Life Sciences DURAN™ Vigreux Column, with 2 standard ground joints, complete, with slide-on glass jacket
Vigreux columns are used for fractional distillation.
Brand: DWK Life Sciences 242408803
Code : A0
Additional Details : Weight : 0.70000kg
Description
Vigreux columns are used in fractionated distillation for improved separation of substances. The slightly downward angled indentations considerably increase the contact surface between the gas phase and the glass, i.e. the cooling surface.
- Tight closure due to standard ground joint
- With slide-on glass jacket
Specifications
Fractional distillation; Laboratory coolers, condensation of gases and vapors | |
Boro 3.3 | |
NS 29/32 |
650 mm | |
1 Ea. | |
Yes |