missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Glass N 8-1 and N 11-1 0 Small Opening Micro Insert

Product Code. 10349870 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10349870 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10349870 Supplier Macherey-Nagel™ Supplier No. 702968.1
From Chemicals to Chromatography
Use promo code 27991
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Clear
Material Glass
Volume (Metric) 0.15 mL
Style For vials with Small opening
Base Style Conical
Quantity 100 Pk.
For Use With (Equipment) Small Opening Vials
Height (Metric) 31 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.