Description
- Available in three sizes for use with various filter sizes and filtration applications
- Constructed of autoclavable borosilicate glass
- For use in glass vacuum filter holder assemblies
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Filtration with glass vacuum filter holder assemblies |
| Product Type | Glass Funnel |