missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Buechner Funnels

Manufactured from borosilicate glass.

Brand:  Duran™ 213415401

Code : A0

Additional Details : Weight : 0.52000kg

 View more versions of this product

Product Code. 10028570

  • £238.97 / Each


In Stock 17
Add to basket

Description

Description

  • Good thermal shock and chemical resistance
  • Features a glass support for membrane and paper filters
  • Typical applications include qualitative inorganic analysis and preparative chemistry
Specifications

Specifications

USP Standard
110mm
Qualitative inorganic analysis and preparative chemistry
280 mm
Buechner Funnel
22mm
1000 mL
Duran Glass, Borosilicate Glass
Videos
Documents