missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GL7 Rat, anti-Human, Clone: GL-7, Proteintech
Rat Monoclonal Antibody
£47.00 - £137.00
Specifications
| Antigen | GL7 |
|---|---|
| Clone | GL-7 |
| Concentration | 0.5 mg/ml |
| Applications | Flow Cytometry |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18035565
|
Proteintech
65261-1-IG-100UG |
100 μg |
£47.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18046865
|
Proteintech
65261-1-IG-500UG |
500 μg |
£137.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GL7 antibody from Proteintech is a rat monoclonal antibody generated with lps activated dba/j mouse b cells of mouse GL7. This antibody recognizes human, mouse antigen. The GL7 antibody has been validated for the following applications: FC analysis.
Specifications
| GL7 | |
| 0.5 mg/ml | |
| Liquid | |
| Human, Mouse | |
| IgM κ | |
| Store at 2°C to 8°C. Stable for one year after shipment. |
| GL-7 | |
| Flow Cytometry | |
| Rat | |
| 104230 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title