missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GL7 Rat, anti-Human, Clone: GL-7, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65261-1-IG-500UG
This item is not returnable.
View return policy
Description
The GL7 antibody from Proteintech is a rat monoclonal antibody generated with lps activated dba/j mouse b cells of mouse GL7. This antibody recognizes human, mouse antigen. The GL7 antibody has been validated for the following applications: FC analysis.
Specifications
| GL7 | |
| GL-7 | |
| Rat | |
| Primary | |
| Human, Mouse | |
| 900 kDa | |
| IgM κ |
| Flow Cytometry | |
| 0.5 mg/ml | |
| 500 μg | |
| 104230 | |
| Store at 2°C to 8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction