missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, GL 45 Thread Neck, GL 32 Thread Side Arm

Product Code. 10445781
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
2000 mL
5000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Aspirator laboratory bottle with screw closure and PTFE key stopcock

Aspirator laboratory bottle with screw closure, complete with PTFE key stopcock and GL 45 red PBT cap.

  • Supplied ready to use
  • DURAN™ glass and PTFE key stopcock outlet
  • Made of virtually inert, chemically resistant DURAN™ glass
  • Fully compatible with all the GL 45 threads and therefore highly versatile
  • Tabulator with GL 32 thread
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
For Use With (Application) Storage and dispensing of liquids and reagents
Material Borosilicate Glass (3.3), Silicone, PTFE
No. per Pack 1/Pk.
Neck Finish Thread Size GL 45
Autoclavable Autoclavable
Diameter (Metric) 101 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'