missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00
Specifications
| Antigen | GIPC1 |
|---|---|
| Dilution | Western Blot 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GIPC1 Polyclonal specifically detects GIPC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GIPC1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| C19orf3chromosome 19 open reading frame 3, GAIP C-terminus-interacting protein, GIPC PDZ domain containing family, member 1, GIPCRGS-GAIP-interacting protein, GLUT1 C-terminal binding protein, GLUT1CBP, IGF-1 receptor interacting protein 1, MGC15889, MGC3774, NIP, PDZ domain-containing protein GIPC1, regulator of G-protein signalling 19 interacting protein 1, RGS19IP1IIP-1, SEMCAP, SYNECTIIN, SYNECTIN, Tax interaction protein 2, TIP-2RGS19-interacting protein 1 | |
| GIPC1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 10755 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EAINGQSLLGCRHYEVARLLKELPRGRTFTLKLTEPRKAFDMISQRSAGGRPGSGPQLGTGR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title