missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Goat Serum, New Zealand origin

Excellent value for basic cell culture, specialty research, and specific assays

Brand:  Gibco™ 16210072

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11540526

  • £338.00 / 500mL
In Stock
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
  • It has earned the trust of researchers with consistent quality and award-winning support
  • Helps meet research needs and budget requirements
  • Shipped frozen, store at <-10°C
  • Sera Category: Specialty
  • Origin: New Zealand
  • Age: Adult
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Goat/Caprine
Liquid
500 mL
Sterile
Gibco™
Adult
New Zealand
Goat Serum
Standard (Sterile-filtered)
Sustainable packaging
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For In Vitro Diagnostic Use. For Research Use Only. Not for use in diagnostic procedures.