missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Goat Serum, New Zealand origin
Excellent value for basic cell culture, specialty research, and specific assays
£81.75 - £338.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Adult |
Species | Goat/Caprine |
Country of Origin | New Zealand |
Form | Liquid |
Description
- Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
- It has earned the trust of researchers with consistent quality and award-winning support
- Helps meet research needs and budget requirements
- Shipped frozen, store at <-10°C
- Sera Category: Specialty
- Origin: New Zealand
- Age: Adult
Order Info
Shipping Condition: Dry ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Goat/Caprine | |
Liquid | |
Standard (Sterile-filtered) | |
Sustainable packaging | |
Frozen |
Adult | |
New Zealand | |
Goat Serum | |
Sterile | |
Gibco™ |
For In Vitro Diagnostic Use. For Research Use Only. Not for use in diagnostic procedures.