missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ghrelin/Obestatin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89773
This item is not returnable.
View return policy
Description
Ghrelin/Obestatin Polyclonal specifically detects Ghrelin/Obestatin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ghrelin/Obestatin | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51738 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GHRL | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RKESKKPPAKLQPRALAGWLRPEDGGQAEGAEDELEVRFNAPFDVGIKLSGVQYQQHS | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction