missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Rat, Unconjugated, Clone: 3H9, Proteintech ChromoTek
Rat Monoclonal Antibody
£137.40 - £178.00
Specifications
| Antigen | GFP |
|---|---|
| Clone | 3H9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Rat monoclonal [3H9] to GFP
Specifications
| GFP | |
| Western Blot | |
| Unconjugated | |
| IgG2a | |
| Shipped at ambient temperature. Store at +4°C. Stable for one year. |
| 3H9 | |
| Monoclonal | |
| Rat | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title