Learn More
GFP Rat, Unconjugated, Clone: 3H9, Proteintech ChromoTek
Rat Monoclonal Antibody
Brand:  Chromotek 3h9-20
 View more versions of this product
Product Code. 17373263
  • £137.40 / 20µL
Description
Rat monoclonal [3H9] to GFP
Specifications
Specifications
GFP
Monoclonal
Unconjugated
20 μL
Shipped at ambient temperature. Store at +4°C. Stable for one year.
Western Blot
3H9
Rat
Primary
IgG2a