missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Rabbit anti-Jellyfish, Clone: 208, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90178-50UL
This item is not returnable.
View return policy
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| GFP | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Jellyfish | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 208 | |
| Western Blot 1:1000-1:5000, ELISA 1:10000-1:20000, Immunoprecipitation 1-4 ul/mg of lysate | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Recombinant Aequorea victoria GFP protein | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction