missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Rabbit anti-Jellyfish, Clone: 208, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £411.00
Specifications
| Antigen | GFP |
|---|---|
| Clone | 208 |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:10000-1:20000, Immunoprecipitation 1-4 ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257047
|
Novus Biologicals
NBP2-90178-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221248
|
Novus Biologicals
NBP2-90178-100UL |
100 μg |
£411.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| GFP | |
| Western Blot 1:1000-1:5000, ELISA 1:10000-1:20000, Immunoprecipitation 1-4 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| Recombinant Aequorea victoria GFP protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 208 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Jellyfish | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| IgG | |
| Protein A purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit