missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£169.00 - £377.00
Specifications
| Antigen | GFP |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18778463
|
Novus Biologicals
NBP2-37821 |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669525
|
Novus Biologicals
NBP2-37821-0.025mg |
0.025 mg |
£169.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| GFP | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Jellyfish | |
| anti-gfp (green fluorescent protein) pab, clone 9F9.F9, Enhanced Green Fluorescent Protein, GFP immunofluorescence, GFP immunoprecipitation, GFP monoclonal, GFP mouse, GFP staining, GFP western blot, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Purified recombinant eGFP | |
| Primary | |
| Detects eGFP in samples. This product has been successfully used in Western Blot and ICC/IF applications. The immunogen is purified recombinant eGFP. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Biologically Active Proteins, Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| PBS (pH 8.4) with No Preservative | |
| gfp | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title