missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-37821
This item is not returnable.
View return policy
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| GFP | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 8.4) with No Preservative | |
| gfp | |
| Purified recombinant eGFP | |
| 100 μg | |
| Biologically Active Proteins, Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| Detects eGFP in samples. This product has been successfully used in Western Blot and ICC/IF applications. The immunogen is purified recombinant eGFP. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:200 | |
| anti-gfp (green fluorescent protein) pab, clone 9F9.F9, Enhanced Green Fluorescent Protein, GFP immunofluorescence, GFP immunoprecipitation, GFP monoclonal, GFP mouse, GFP staining, GFP western blot, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Jellyfish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction