missing translation for 'onlineSavingsMsg'
Learn More

Gestonorone, TRC

Product Code. 30396992
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396992 2.5 g 2.5g
30391559 1 g -
30400257 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396992 Supplier Toronto Research Chemicals Supplier No. TRCG3682702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2137-18-0
Molecular Formula C20 H28 O3
Molecular Weight (g/mol) 316.43
Synonym 19-Norpregn-4-ene-3,20-dione, 17-hydroxy-, 17-Hydroxy-19-norpregn-4-ene-3,20-dione, 17α-17-Acetyl-17-hydroxyestr-4-en-3-one, 17α-Hydroxy-19-norpregn-4-en-3,20-dione, 17α-Hydroxy-19-norpregn-4-ene-3,20-dione, 17α-Hydroxy-19-norprogesterone, 17β-Acetyl-17α-hydroxyestr-4-en-3-one, 19-Norpregn-4-en-3,20-dione-17α-ol, Gestonorone, Gestronol, TX 045
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C20H28O3/c1-12(21)20(23)10-8-18-17-5-3-13-11-14(22)4-6-15(13)16(17)7-9-19(18,20)2/h11,15-18,23H,3-10H2,1-2H3/t15-,16+,17+,18-,19-,20-/m0/s1
Formula Weight 316.2038
Chemical Name or Material Gestonorone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.