CAS RN 2137-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gestonorone, TRC
CAS: 2137-18-0 Molecular Formula: C20 H28 O3 Molecular Weight (g/mol): 316.43 Synonym: 19-Norpregn-4-ene-3,20-dione, 17-hydroxy-,17-Hydroxy-19-norpregn-4-ene-3,20-dione,17α-17-Acetyl-17-hydroxyestr-4-en-3-one,17α-Hydroxy-19-norpregn-4-en-3,20-dione,17α-Hydroxy-19-norpregn-4-ene-3,20-dione,17α-Hydroxy-19-norprogesterone,17β-Acetyl-17α-hydroxyestr-4-en-3-one,19-Norpregn-4-en-3,20-dione-17α-ol,Gestonorone,Gestronol,TX 045 IUPAC Name: (8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one SMILES: CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C