missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ TRα-UAS-bla HEK 293T Assay Kit

Product Code. 10318763
Click to view available options
Quantity:
1 Plate
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10318763

Brand: Thermo Scientific™ K1684

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Thyroid hormone receptor alpha(TR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.

TR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Thyroid hormone receptor alpha(TR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-TR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. TR alpha-UASbla HEK 293T 293 cells are functionally validated for Z' and EC50 concentrations of T3 Thryoid hormone. In addition, TR alpha-UAS-bla HEK 293T 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists 9-cis-retinoic acid, TTNPB, CD437, AM 580
Antagonists AGN 193109
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage TR alpha-UAS-bla HEK 293T cells (K1260)

Shipping: Dry Ice
Storage:-80°C (liquid nitrogen)
Description Primary Agonist: All-trans-retinoic Acid; Antagonists: AGN 193109; Reporter: BLA (Beta-Lactamase)
Detection Method Fluorescence
For Use With (Application) Nuclear Receptors
Includes TR alpha-UAS-bla HEK 293T cells (K1260)
Primary Agonist All-trans-retinoic Acid
Product Line GeneBLAzer
Quantity 1 Plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry THRA, TR alpha, NR1A1
Type Detection Assay Kit
Cell Line HEK 293T
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.