Learn More
Thermo Scientific™ GeneBLAzer™ RAR-gamma DA Assay Kit
Description
GeneBLAzer™RAR gamma DA (Division Arrested) cells and RAR gamma-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human retinoic acid receptor gamma (RAR gamma) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-RAR gamma (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both RAR gamma DA cells and RAR gamma-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of all-trans retinoic acid (ATRA). In addition, RAR gamma-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and stimulation time.
Specifications
Specifications
| Alternate Agonists | 4-hydroxytamoxifen, Chlordane, Toxaphene |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Division-Arrested Cells |
| Content And Storage | Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate. • RAR gamma HEK 293T DA cells (K1559A) • LiveBLAzer™-FRET B/G Loading Kit, 70 μg Shipping: Dry Ice Storage: -80°C (liquid nitrogen) |
| Description | GeneBLAzer™ RAR-gamma DA Assay Kit, 1 Plate, ∽2,400,000 cells/vial Quantity of Cells, 293T (HEK) Cell Line, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | Pharma and biopharma, target-based adme/tox assays, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays |
| Gene Symbol | RARγ |
| Includes | RAR gamma HEK 293T DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg |
| Primary Agonist | XCT790 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.