Learn More
Invitrogen™ GeneBLAzer™ PPAR delta DA Assay Kit
GeneBLAzer™ PPAR delta DA Assay Kit
Brand: Invitrogen™ K1395
Code : NEW
Additional Details : Weight : 0.04000kg
Description
GeneBLAzer™ PPAR delta DA (Division Arrested) cells and PPAR delta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-delta (PPAR delta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line. GeneBLAzer™ UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-PPAR delta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both PPAR delta DA cells and PPAR delta-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of L-165041. In addition, PPAR delta-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.Specifications
Liquid Nitrogen | |
Fluorescent | |
PPAR delta HEK 293T DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg | |
293T (HEK) |
GeneBLAzer™ PPAR delta DA Assay Kit, 1 Plate, ∽4,800,000 cells/vial at 4,800,000 cells/mL Quantity of Cells, 293T (HEK) Cell Line, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage | |
Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays, Cell analysis, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, nuclear receptor biology, cell-based nuclear receptor reporter assays, target-based ADME/tox assays, cellular adme assays, nuclear receptor cellular adme assays | |
1 Plate | |
Division-Arrested Cells Assay Kit |
For Research Use Only. Not for use in diagnostic procedures.