missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ OPRK1-Gqo5-NFAT-bla CHO-K1 Cells

Product Code. 10782004
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10782004

Brand: Thermo Scientific™ K1533

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K1 cells contain the human Opioid Receptor Kappa 1 (OPRK1) (Accession No. NM_000710) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line.

The GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K1 cells contain the human Opioid Receptor Kappa 1 (OPRK1) (Accession No. NM_000710) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. The GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells (Cat. No. K1536) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element and the chimeric G Protein, Gqo5, stably integrated into CHO-K1 cells.

The GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K11 cells are functionally validated for Z'-factor and EC50 concentrations using U50488. In addition, GeneBLAzer™ OPRK-1-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000912
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ OPRK1 Gqo5 NFAT-bla CHO-K1 cells
• GeneBLAzer™ OPRK1 Gqo5 NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ OPRK1 Gqo5 NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol OPRK1
Includes OPRK1 Gqo5 NFAT-bla CHO-K1 cells, OPRK1 Gqo5 NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry OPRK1
Type CHO-K1 Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.