missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ Jurkat Control Kit

Product Code. 10207912
Click to view available options
Quantity:
1 kit
Unit Size:
2mL
This item is not returnable. View return policy

Product Code. 10207912

Brand: Thermo Scientific™ K1206

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The GeneBLAzer™ Jurkat Control Kit contains constitutively expressing (CMV-bla) and wildtype Jurkat cells to provide positive and negative controls, respectively, for evaluating beta-lactamase activity (Figure 1).

The GeneBLAzer™ Jurkat Control Kit contains constitutively expressing (CMV-bla) and wildtype Jurkat cells to provide positive and negative controls, respectively, for evaluating beta-lactamase activity (Figure 1).

TRUSTED_SUSTAINABILITY

Specifications

Cell State Dividing Cells
Content And Storage The GeneBLAzer™ Jurkat Control Kit contains 2 x 1 mL vials of cell s(∼5 x 106 cells/mL) and is shipped on dry ice. Store in liquid nitrogen upon receipt.
Description GeneBLAzer™ Jurkat Control Kit, 1 Kit, ∽ 5,000,000 cells/vial at 5,000,000 cells/mL Quantity of Cells, Jurkat Cell Line, Fluorescent Detection Method, GPCRs, Signaling Pathway Druggable Target, Suspension Growth Properties, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Cell analysis, cellular imaging, cell-based reporter assays, cell-based and beta-Lactamase assays
Includes Two vials of cells, onemL per vial (∽5 x 106 cells/mL)
Product Line GeneBLAzer
Quantity 1 kit
Sample Type Jurkat
Type Cell Line
Cell Line Jurkat
System Type GeneBLAzer™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.