missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ Gqo5-NFAT-bla FreeStyle™ 293F Cells

Product Code. 10379283
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10379283

Brand: Thermo Scientific™ K1747

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ Gqo5-NFAT-bla FreeStyle™ 293F cells contain the chimeric G-protein Gqo5 stably integrated into the CellSensor™ NFAT-blaFreeStyle™ 293F cell line.

GeneBLAzer™ Gqo5-NFAT-bla FreeStyle™ 293F cells contain the chimeric G-protein Gqo5 stably integrated into the CellSensor™ NFAT-blaFreeStyle™ 293F cell line. CellSensor™ NFAT-bla FreeStyle™ 293F cells contain a beta-lactamase reporter gene under control of a stably integrated NFAT response element. Gqo5-NFAT-bla FreeStyle™ 293F is a parental cell line designed to be used for the construction of Gi/o-coupled GPCR beta-lactamase cellular assays. The Gi/o receptor of interest should be stably expressed in the cell line by expression from the selectable vector pcDNA™6.2-hygro-DEST (Cat. No. K1233). Gqo5-NFAT-bla FreeStyle™ 293F cells have been shown to respond to the agonist of a transiently expressed D2 receptor.

TRUSTED_SUSTAINABILITY

Specifications

Cell State Dividing Cells
Content And Storage Shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) GPCRs
Gene Symbol Gqo5
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Type Dividing Cells
Cell Line FreeStyle 293-F Cells
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.