Learn More
Thermo Scientific™ GeneBLAzer™ ERRα-UAS-bla HEK 293T Cells
Description
ERR alpha-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Estrogen related receptor alpha (ERR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-ERR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. ERR alpha-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of XCT790, an inverse agonist. In addition, ERR alpha-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
Specifications
Specifications
| Alternate Agonists | L165, 041, GW 1929, GW 7647, GW 0742 |
| Antagonists | T0070907, GW 9662, GW 6471 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | GeneBLAzer™ Nuclear Receptor Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use., Liquid Nitrogen |
| Detection Method | Fluorescence, Fluorescent |
| For Use With (Application) | Nuclear Receptors |
| Druggable Target | Nuclear Receptors |
| No. of Cells | 2 x 106 Cells |
| Primary Agonist | Rosiglitazone |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.