missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ CMV-bla Jurkat Positive Control Cell Line

Product Code. 10686493
Click to view available options
Quantity:
1 vial
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10686493

Brand: Thermo Scientific™ K1045

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The GeneBLAzer™ CMV-bla Jurkat Cell Line constitutively expresses beta-lactamase under the control of the CMV promoter in Jurkat cells.

A stable population was obtained using 1 mg/ml Geneticin™ (G418) selection followed by isolation of a clone by fluorescence-activated cell sorting (FACS).

CMV-bla Jurkat cells can be used as a positive control for beta-lactamase expression experiments. When the cells are loaded with CCF2-AM or CCF4-AM substrate and excited with 409 nm light, the cells will fluoresce blue.

TRUSTED_SUSTAINABILITY

Specifications

Cell State Dividing Cells
Content And Storage GeneBLAzer™ CMV-bla Jurkat Positive Control Cell Line is shipped on dry ice and should be stored in liquid nitrogen.
Detection Method Fluorescence
Product Line GeneBLAzer™
Quantity 1 vial
Shipping Condition Dry Ice
Type Cell Line
Cell Line Jurkat
System Type CellSensor™
Druggable Target GPCR, Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato