missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GeneBLAzer™ CHO-K1 Control Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
2mL
Description
The GeneBLAzer™ CHO-K1 Control Kit contains constitutively expressing (CMV-bla) CHO-K1 cells and wildtype CHO-K1 cells to provide positive and negative controls for evaluating beta-lactamase activity.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | The GeneBLAzer™ CHO-K1 Control Kit contains 2 x 1 mL vials of cells (∼3 x 106 cells/mL for CMV-bla CHO-K1 and ∼2 x 106 cells/mL for CHO-K1) and is shipped on dry ice. Store in liquid nitrogen upon receipt. |
| Description | GeneBLAzer™ CHO-K1 Control Kit, 1 Kit, ∽ 2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs, Signaling Pathway Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell analysis, cellular imaging, cell-based reporter assays, cell-based and beta-Lactamase assays |
| Includes | Two vials of cells, onemL per vial (∽3 x 106 cells/mL for CMV-bla CHO-K1 and ∽2 x 106 cells/mL for CHO-K1) |
| Product Line | GeneBLAzer |
| Quantity | 1 Kit |
| Sample Type | CHO-K1 |
| Type | CHO-K1 Control Kit |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction