Learn More
Agilent BioTek Gen6 Data Analysis Software
GREENER_CHOICE
Product Code. 17864677 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Microplate Reader
Unit Size:
Each
Product Code. 17864677
missing translation for 'mfr': Agilent BioTek GEN6
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Gen6 software provides full control of Agilent BioTek multimode and absorbance readers for plate reading in 6- to 384-well microplates, for both endpoint and kinetic assays. Reading parameters, plate layout and data analysis definition is an easy, step-by-step process for quantitative and qualitative assays.
  • Walk-up usability
    • Both new and experienced users will be able to walk up and use Gen6 without extensive training required.
  • Set up assay, analyze results, and export data in minutes
    • With Gen6, users can set up their plate, read, and analyze results in minutes.
  • Prebuilt experiment templates in Gen6
    • Prebuilt experiment templates allow new users to quickly and easily get started with common assays and applications within Gen6.
  • Advanced read functionality including kinetics and area scans
    • Gen6 contains advanced functionality for kinetic assays. Users can easily set up time-course experiments and visualize data in real-time as the microplate reader collects it. Raw and calculated data can easily be exported with one-click into Microsoft Excel.
  • Advanced reading modes, such as area scans, allow users to measure discrete points across a well for heterogeneous samples.
  • Instrument compatibility: Synergy Neo2, Synergy H1, Synergy HTX, Synergy LX, Epoch 2, Epoch, and 800 TS
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Microplate Reader
Type Software