missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Gen5 Spot Counting Module Add-On

Product Code. 16245592 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Gen5 Image Prime
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16245592

Brand: Agilent BioTek GEN5SPOT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gen5 Spot Counting Module Add-On for Gen5 Image Prime only. Requires Gen5 Image Prime 3.03 or higher.

  • Powerful functionality microplate reading, enabling applications from micro-volume nucleic acid quantification to long-term cell-based kinetic assays.
  • Fluorophore spectra utility offers >100 fluorophore spectra, making filter cube matching quick and easy
  • Enhanced conditional logic adds powerful functionality for data transformation, cutoffs and validation with automated outlier rejection and expanded formula operators
  • Improved file size, sorting and search tools for working with Gen5's database and sample file database
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Gen5 Image Prime
Type Software
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.