Learn More
Aligent™ BioTek™ Gen5 Object Tracking Module
GREENER_CHOICE
Product Code. 17864667 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Gen5
Unit Size:
Each
Product Code. 17864667
Brand: Aligent™ BioTek™ GEN5TRACK
The Agilent BioTek™ Gen5 Object Tracking Module provides an automated method for characterizing 2D cell migration in high-content formats, including 96-well microplates and chemotaxis slides. Gen5 Image Prime software is required.
The concerted movement of cells within their environment plays a fundamental role in nearly all eukaryotic organisms. Higher eukaryotes rely on regulated cell migration throughout development and for immunologic processes while aberrant cell migration is a defining feature of metastatic cancers. The process of cellular migration towards external stimuli (such as chemotaxis) is called directional cell migration. Equally as informative is random cell migration in which cell trajectories are measured when a chemoattractant is diffuse instead of emanating from a source direction. Whether studying directional or random migration it is important to measure the motile capacity of cells accurately and reliably.
  • Gen5 Object Tracking Module provides an automated method for characterizing 2D cell migration in high-content formats
  • Combined with Agilent™ BioTek™ automated imaging systems, this capability delivers an integrated solution for automating cell migration studies
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Characterizing 2D Cell Migration
Required Accessories Gen5 Image Prime Software
For Use With (Equipment) Gen5
Type Software