missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemin 7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58154-25ul
This item is not returnable.
View return policy
Description
Gemin 7 Polyclonal specifically detects Gemin 7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Gemin 7 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| FLJ13956, gem (nuclear organelle) associated protein 7, gem-associated protein 7, gemin 7, Gemin-7, SIP3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79760 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| GEMIN7 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GPDGFSRGFAPDGRRAPLRPEVPEIQECPIAQESLESQEQRARAALRERYLRSLLAM | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering