missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £470.00
Specifications
| Antigen | Gemin 3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18458600
|
Novus Biologicals
NBP1-84058-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18255288
|
Novus Biologicals
NBP1-84058 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Gemin 3 Polyclonal specifically detects Gemin 3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| Gemin 3 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 11218 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MAKLKHFHCRVLISTDLTSRGIDAEKVNLVVNLDVPLDWETYMHRIGRAGRFGTLGLTVTYCCRGEEENMMMRIAQKCNINLLPLPDPIPSGLMEECVDWDVEVKAAVHTYGIASVPNQPLKKQIQKIERTLQIQKAHGDH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Component of gems 3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 20, DEAD box protein 20, DEAD box protein DP 103, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD, DKFZp434H052, DP103DEAD-box protein DP103, EC 3.6.4.13, Gemin-3, GEMIN3gemin-3, probable ATP-dependent RNA helicase DDX20, SMN-interacting protein | |
| DDX20 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts