missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Gel Casting Tray

Forms leak proof seal against inner walls.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10142041

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Silicone gaskets to line casting tray
  • Forms leak proof seal against inner walls of running chamber when casting tray is turned at 90° to direction of electrophoresis
Specifications

Specifications

Gel Casting Tray
Acrylic
200 mm
Leakproof Silicone Seals
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers