Learn More
Fisherbrand™ Gel Casting Tray
Forms leak proof seal against inner walls.
£147.00 - £419.00
Product Code | Brand | Length (Metric) Plate | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Plate | Width (Metric) | Price | Quantity & Availability | ||||
10142041
|
Fisherbrand™
|
200 mm | 200 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10569591
|
Fisherbrand™
|
130 mm | 128 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11709665
|
Fisherbrand™
|
300 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10395541
|
Fisherbrand™
|
75 mm | 60 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10479352
|
Fisherbrand™
|
115 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Accessory for horozontal gel electrophoresis units
- Silicone gaskets to line casting tray
- Forms leak proof seal against inner walls of running chamber when casting tray is turned at 90° to direction of electrophoresis
Specifications
Gel Casting Tray | |
Acrylic |